Anti-KLKL2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX22291
89361-210EA 395.06 USD
89361-210
Anti-KLKL2 Rabbit Polyclonal Antibody
Antibodies
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L2 is expressed in breast, brain, and testis and is up-regulated by estrogens and progestins.

For WB: Use at a dilution of 1:500 - 1:1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: KLKL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR