Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex
GTX27152
89362-458EA 247.41 USD
89362-458
Anti-IgG Goat Polyclonal Antibody
Antibodies
Biotin-conjugated Goat anti-Human IgG polyclonal antibody, pre-adsorbed

Recommended Starting Dilutions:Dot: Use at an assay dependant dilution. ELISA: Use at a dilution of 1/20000 - 1/80000. IHC-P: Use at an assay dependant dilution.IHC-Fr: Use at an assay dependant dilution. IM: Use at an assay dependant dilution. WB: Use at an assay dependant dilution.Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR