Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MUC13 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX87168
89366-918EA 398.53 USD InStock
89366-918
Anti-MUC13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MUC13

We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 IF: 1:100~1:500 ELISA: 1:5000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty