Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 17F5]

Supplier: GeneTex
GTX54143
89363-708EA 587.47 USD
89363-708
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 17F5]
Antibodies
Mouse monoclonal antibody [17F5] to MAdCAM-1

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/100 Immunohistology - Paraffin: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: MADCAM1
Clonality: Monoclonal
Clone: 17F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR