Anti-IRS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24873
89361-570EA 611.89 USD
89361-570
Anti-IRS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to IRS1 ( phospho Y896 )

WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Predicted molecular weight: 165 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR