Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgE Sheep Polyclonal Antibody

Supplier: Genetex

GTX77500
89353-748EA 167.59 USD InStock
89353-748
Anti-IgE Sheep Polyclonal Antibody
Antibodies
HRP-conjugated Sheep anti-Human IgE polyclonal antibody

Recommended Starting Dilutions:
For ELISA: Use at a concentration of 0.1 - 0.01 ug/ml.
For WB: Use at a concentration of 0.5 - 0.05 ug/ml.
For IHC: Use at a concentration of 5.0 - 0.5 ug/ml.
Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty