Anti-IgA Goat Polyclonal Antibody

Supplier: GeneTex
GTX29157
89352-580EA 236.75 USD
89352-580
Anti-IgA Goat Polyclonal Antibody
Antibodies
Goat anti-Human IgA polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR