Anti-HTR4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77686
89366-238EA 610.74 USD
89366-238
Anti-HTR4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to 5-HT4 receptor

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: HTR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR