Anti-HSP70 1A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX104126
89350-248EA 427.1 USD
89350-248
Anti-HSP70 1A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HSP70 1A (heat shock 70kDa protein 1A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:500-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HSP70 1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR