Anti-HSP27 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX17937
89360-350EA 587.58 USD
89360-350
Anti-HSP27 Rabbit Polyclonal Antibody
Antibodies
Phosphorylation of HSP27 on serine 82 by MAPKAP-kinase 2 leads to HSP27 dissociation from the Akt/MAPKAP-kinase 2/p38 complex and from actin filaments, and stimulates HSP27 binding to the IKK complex.

WB: 1/1000. Detects a band of approximately 27 kDa (predicted molecular weight: 27 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HSP27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR