Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP20 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX13491
89359-688EA 405.71 USD InStock
89359-688
Anti-HSP20 Rabbit Polyclonal Antibody
Antibodies
Hsp20 is a small heat shock protein related to Hsp25, Hsp27 and may form different hetercomplexes with these proteins. The specific physiological function of Hsp20 is not yet known. It is distributed ubiquitously in tissues, but is found in higher levels in skeletal, smooth and heart muscle. Under normal conditions, Hsp20 is diffusely distributed in the cytosol, but under heat stress conditions, it translocates to the nucleus. Unlike other heat shock proteins the amount of Hsp20 does not increase after heat shock.

WB: Use at a concentration of 0.05 μg/ml. Detects a band of approximately 20 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HSP20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty