Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/2]

Supplier: GeneTex
GTX78394
89366-624EA 614.11 USD
89366-624
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/2]
Antibodies
Monoclonal Antibody to HLA-G (Human) Purified

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR