To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:HLA class I histocompatibility antigen, E alpha chain
- Antigen Symbol:HLA-E
- Clonality:Monoclonal
- Clone:MEM-E/02
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3133
- Antigen Synonyms:EA2.1|HLA class I histocompatibility antigen|HLA-6.2|lymphocyte antigen|MHC HLA-E alpha-1|E alpha chain|DAMA-277I14.1|DKFZp686P19218|MHC class I antigen E|EA1.2|MHC|alpha chain E|QA1|MHC HLA-E alpha-2.1
- Storage Buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant denatured heavy chain (Human).
- Tested Applications:IHC-P
- Purification:>95% by SDS-PAGE
- Cat. No.:89361-186
- Supplier no.:GTX22216
HLA E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA E binds a restricted subset of peptides derived from the leader peptides of other class I molecules.
IHC-P: Use at an assay dependant dilution. WB: Use at an assay dependant dilution. Predicted molecular weight: 43 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: HLA-E
Clonality: Monoclonal
Clone: MEM-E/02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human