Anti-HLA-DR1 Mouse Monoclonal Antibody [clone: MEM-267]

Supplier: GeneTex
GTX78393
89366-622EA 623.54 USD
89366-622
Anti-HLA-DR1 Mouse Monoclonal Antibody [clone: MEM-267]
Antibodies
Type: Primary
Antigen: HLA-DR1
Clonality: Monoclonal
Clone: MEM-267
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR