Anti-HLA-A2 Mouse Monoclonal Antibody [clone: BB7.2]

Supplier: GeneTex
GTX75803
89365-912EA 592.7 USD
89365-912
Anti-HLA-A2 Mouse Monoclonal Antibody [clone: BB7.2]
Antibodies
Mouse monoclonal antibody [BB7.2] to HLA-A2

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 10 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HLA-A2
Clonality: Monoclonal
Clone: BB7.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR