Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIV1 GP41 Mouse Monoclonal Antibody [clone: 5A1]

Supplier: Genetex

GTX19975
89367-516EA 486.95 USD InStock
89367-516
Anti-HIV1 GP41 Mouse Monoclonal Antibody [clone: 5A1]
Antibodies
The antibody may be used to detect HIV1 envelope glycoprotein in virus or infected cell preparations.

ELISA: Use at an assay dependent dilution. WB: 1/1000. Is unsuitable for IHC. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HIV1 GP41
Clonality: Monoclonal
Clone: 5A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty