Anti-H3 Mouse Monoclonal Antibody [clone: 117C826]

Supplier: GeneTex
GTX13924
89359-910EA 626.05 USD
89359-910
Anti-H3 Mouse Monoclonal Antibody [clone: 117C826]
Antibodies
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fibre is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures.

WB: Use at a concentration of 2 ?g/ml. Detects a band of approximately 18 kDa (predicted molecular weight: 16.3 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. We advise you to centrifuge this product vial before use.

Type: Primary
Antigen: H3F3A
Clonality: Monoclonal
Clone: 117C826
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR