Anti-Histidine TAG Mouse Monoclonal Antibody

Supplier: GeneTex
GTX40628
89367-714EA 565.51 USD
89367-714
Anti-Histidine TAG Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Histidine TAG
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR