Anti-MET Goat Polyclonal Antibody

Supplier: GeneTex
GTX10728
89354-896EA 610.36 USD
89354-896
Anti-MET Goat Polyclonal Antibody
Antibodies
Goat polyclonal to Met (c-Met)

ELISA: Use at a concentration of 0.5 - 1.0 ?g/ml. Neut: Use at a concentration of 0.5 - 2.0 ?g/ml. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 129 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR