Anti-Hepatitis C NS5B Mouse Monoclonal Antibody

Supplier: GeneTex
GTX30873
89363-010EA 567.41 USD
89363-010
Anti-Hepatitis C NS5B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis C NS5B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR