Anti-HBeAg Mouse Monoclonal Antibody

Supplier: GeneTex
GTX42062
89363-490EA 769.58 USD
89363-490
Anti-HBeAg Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis B 'E' Antigen (HBEAG)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR