Anti-HEC1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX23393
89361-354EA 577.91 USD
89361-354
Anti-HEC1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to HEC1

  • Approx 75-80kDa band observed in Hela lysate (predicted MW of 74kDa according to NP_006092). Recommended for use at 0.5-2µg/ml.

Type: Primary
Antigen: HEC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR