Anti-HSP70 Mouse Monoclonal Antibody [clone: 3A3]

Supplier: GeneTex
GTX25439
89367-594EA 604.24 USD
89367-594
Anti-HSP70 Mouse Monoclonal Antibody [clone: 3A3]
Antibodies
Mouse monoclonal antibody [3A3] to HSP70

Recommended Starting Dilutions:IHC-P: 1:200ICC/IF: 1:50 - 1:200ELISA: 1:500FACS: 1:100 - 1:200IHC: 1:1000WB: 1:100 - 1:5000IP: 2 µlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Amphibian
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR