Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-HDL Chicken Polyclonal Antibody

Supplier: Genetex

GTX15863
89367-446EA 457.73 USD InStock
89367-446
Anti-HDL Chicken Polyclonal Antibody
Antibodies
Cholesterol-carrier protein that acts as scavenger of tissue cholesterol. Important in cholesterol efflux from tissues. Involved in return of cholesterol from the periphery to the liver for removal as bile acids.

Type: Primary
Antigen: HDL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty