Anti-HA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30574
89367-092EA 839.46 USD
89367-092
Anti-HA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR