Anti-HA Chicken Polyclonal Antibody

Supplier: GeneTex
GTX77397
89353-682EA 601.14 USD
89353-682
Anti-HA Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR