Anti-GST Goat Polyclonal Antibody

Supplier: GeneTex
GTX26635
89361-978EA 878.31 USD
89361-978
Anti-GST Goat Polyclonal Antibody
Antibodies
This highly active polyclonal antibody is directed against purified recombinant Glutathione-S-Transferase (GST) cloned from the organism Schistosoma japonicum. This reagent is specifically designed for screening and recovery of fusion proteins from expression systems using a GST gene cloning site

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR