Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111192
89356-624EA 582.76 USD
89356-624
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GSK3 beta (glycogen synthase kinase 3 beta)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR