Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX86837
89366-906EA 616.68 USD
89366-906
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GSK3 beta (phospho Ser9)

We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 ELISA: 1:5000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR