Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSF3R Mouse Monoclonal Antibody

Supplier: Genetex

GTX11411
89357-892EA 367.92 USD InStock
89357-892
Anti-GSF3R Mouse Monoclonal Antibody
Antibodies
Granulocyte colony-stimulating factors promotes survival, proliferation and differentiation of bone marrow precursor cells. GCSF is a lineage-restricted hematopoietic growth factor, stimulating final mitotic divisions and terminal cellular maturation of partially differentiated hematopoietic progenitors.

Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: LMM775
Conjugation: Phycoerythrin (PE)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty