Anti-GFP Rabbit Polyclonal Antibody

GTX77383
89353-674EA 403.61 USD
89353-674
Anti-GFP Rabbit Polyclonal Antibody
Type: Primary
Antigen: Green Fluorescent
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR