Anti-GORASP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112112
89356-982EA 585.66 USD
89356-982
Anti-GORASP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GRASP65 (golgi reassembly stacking protein 1, 65kDa)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: GORASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR