Anti-GP200 Mouse Monoclonal Antibody [clone: PN-15]

GTX72343
89353-000EA 341.65 USD
89353-000
Anti-GP200 Mouse Monoclonal Antibody [clone: PN-15]
Mouse monoclonal [PN-15] to Renal Cell Carcinoma Marker (Gp200)

For WB: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: PN-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR