Anti-GOT2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX88925
89366-968EA 607.87 USD
89366-968
Anti-GOT2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to GOT2 (aa 360 to 373)

Approx 48kDa band observed in Human Liver lysates (calculated MW of 47.5kDa according to NP_002071.2). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: GOT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR