Anti-GMCSF1 Rat Monoclonal Antibody [clone: MP1-22E9]

Supplier: GeneTex
GTX13788
89359-854EA 476.03 USD
89359-854
Anti-GMCSF1 Rat Monoclonal Antibody [clone: MP1-22E9]
Antibodies
This cytokine stimulates the growth and differentiation of hematopoietic precursor cells from various lineages, including granulocytes, macrophages, eosinophils and erythrocytes. Used in myeloid reconstitution following bone marrow transplant, bone marrow transplant engraftment failure or delay, mobilization and following transplantation of autologous peripheral blood progenitor cells, and following induction chemotherapy in older adults with acute myelogenous leukemia.

Recommended Starting Dilutions: For ELISA: Use at an assay dependent dilution. For ELISPOT: Use at an assay dependent dilution. For IHC-Fr: Use at an assay dependent dilution on GM-CSF expressing mouse cells. For WB: Use at a concentration of 1 - 5 ?g/ml, this concentration can detect less than 100 ng of recombinant mouse GM-CSF under reducing conditions. Not tested in other applications. Optimal dilution should be determined by the end user.

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: MP1-22E9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR