Anti-GMCSF1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10669
89354-758EA 611.65 USD
89354-758
Anti-GMCSF1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to GM - CSF

ELISA: Use at a concentration of 0.5 - 1.0 ?g/ml to detect a limit of ~0.6 ng/well of recombinant rat GM-CSF. Neut: Use at a concentration of 0.3 - 0.6 ?g/ml to neutralise 50% of the bioactivity due to 0.5 ng/ml recombinant rat GM-CSF using DA3 cells. WB: Use at a concentration of 0.1 - 0.2 ?g/ml to detect rat GM-CSF at 1 ng/lane under non-reducing and reducing conditions. Detects a band of approximately 22 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR