Anti-GM2A Rabbit Polyclonal Antibody

GTX101184
89348-876EA 176.03 USD
89348-876
Anti-GM2A Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to GM2A (GM2 ganglioside activator)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: GM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR