Anti-GK Goat Polyclonal Antibody

Supplier: GeneTex
GTX40600
89363-258EA 788.34 USD
89363-258
Anti-GK Goat Polyclonal Antibody
Antibodies
Glycerol kinase catalyzes the formation of glycerol 3-phosphate from ATP and glycerol. Dihydroxyacetone and L-glyceraldehyde can also act as acceptors; UTP and, in the case of the yeast enzyme, ITP and GTP can act as donors. It provides a way for glycerol derived from fats or glycerides to enter the glycolytic pathway.

Type: Primary
Antigen: Glycerol Kinase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR