Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIA1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX31031
89363-020EA 382.57 USD InStock
89363-020
Anti-GRIA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Glutamate receptor 1

We recommend the following starting dilutions: Western Blot 1:500-1:1000, immunohistochemistry 1:50-1:200, immunofluorescence 1:50-1:200,. Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: GRIA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty