Anti-GJB1 Mouse Monoclonal Antibody [clone: CXN-32]

Supplier: GeneTex

GTX11366
89357-674EA 428.97 USD
89357-674
Anti-GJB1 Mouse Monoclonal Antibody [clone: CXN-32]
Antibodies
Mouse monoclonal [CXN - 32] to GJB1

ELISA: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000. Detects a band of approximately 27 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GJB1
Clonality: Monoclonal
Clone: CXN-32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR