Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GH1 Mouse Monoclonal Antibody

Supplier: Genetex

GTX27905
89362-684EA 359.27 USD InStock
89362-684
Anti-GH1 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody [GH-45] to Growth Hormone

ICC: Use at an assay dependant dilution. IHC-P: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GH1
Clonality: Monoclonal
Clone: GH-45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty