Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]

Supplier: GeneTex
GTX73615
89353-230EA 618.09 USD
89353-230
Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]
Antibodies
Monoclonal Mouse Antibody to Porcine Glial Fibrillary Acidic Protein ( GFAP )

This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Suitable for formalin/paraffin tissues. Can also be used for: Immunoprecipitation (1:50), Western Blotting (1:50 to 1:100)

Type: Primary
Antigen: Gfap
Clonality: Monoclonal
Clone: GA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR