Rabbit Anti-Gerbil IgG Polyclonal Antibody

GTX77587
89353-806EA 154 USD
89353-806
Rabbit Anti-Gerbil IgG Polyclonal Antibody
Rabbits were immunized with highly purified gerbil IgG isolated from normal gerbil serum. The resulting anti-IgG was immunoaffinity purified off a gerbil IgG immunoabsorbant and conjugated with fluorescein isothiocyanate (FITC).

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Gerbil
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR