Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Anti-Gerbil IgG Polyclonal Antibody

Supplier: Genetex

GTX77589
89353-810EA 145.24 USD InStock
89353-810
Rabbit Anti-Gerbil IgG Polyclonal Antibody
Antibodies
By immunoelectrophoresis and double diffusion, the affinity purified antibodies form a precipitin arc against the IgG in gerbil serum. Rabbits were immunized with gerbil IgG intact molecule. After multiple immunizations in Freund's adjuvant, the rabbits were bled and the serum collected. The specific antibodies were immunoaffinity purified off an IgG affinity column

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Gerbil

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty