Anti-CXCL6 Rabbit Polyclonal Antibody

GTX108446
89355-522EA 176.03 USD
89355-522
Anti-CXCL6 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to GCP2 (chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2))

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CXCL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR