Anti-GAST Rabbit Polyclonal Antibody

GTX16035
89351-206EA 385.9 USD
89351-206
Anti-GAST Rabbit Polyclonal Antibody
This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract,and in the CNS. Antibodies to gastrin are also useful for the identificationand detection of gastrin in normal and neoplastic tissue.

IHC-P: Use at a dilution of 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GAST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Dog, Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR