Anti-GAL7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10482
89350-772EA 610 USD
89350-772
Anti-GAL7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Galectin 7

IP: Use at a concentration of 10 - 20 ?g/mg of lysate. WB: Use at a dilution of 1/10000 - 1/40000. Detects a band of approximately 14 kDa (predicted molecular weight: 16 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LGALS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR