Anti-GADD153 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10444
89350-336EA 609.75 USD
89350-336
Anti-GADD153 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DDIT3

WB: a working dilution of 1:200-1:400 is determined using a whole extract of HEK-293 over-expressing GADD153 and a chemiluminescent detection system.

ICC/IF: a working dilution of 5-10 ug/ml is determined using Hela human cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DDIT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR