Anti-GABAAR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX31012
89363-016EA 674.34 USD
89363-016
Anti-GABAAR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GABAA Receptor, g2 (phospho Ser327)

Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: Gabrg2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR