Anti-GNA15 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX44501
89363-540EA 660.25 USD
89363-540
Anti-GNA15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to G protein alpha 16

For WB: Use at a dilution of 1:4,500 - 1:5,500. The use of this product can also be used for IP but it is not yet reported. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: GNA15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR